Order Entry
Canada
Orders LinkContactUsLinkComponent
Rat Recombinant CNTF (from <i>E. coli</i>)

Rat Recombinant CNTF (from E. coli)

Supplier:   Prosci
Rat Recombinant CNTF (from <i>E. coli</i>)
Supplier:  Prosci
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CNTF is a potent neural factor that was originally characterized as a vital factor for the survival of chick ciliary neurons in vitro. CNTF is also important for the survival of other neural cell types including primary sensory neurons, motor neurons, basal forebrain neurons and type 2 astrocytes. CNTF is highly conserved across species and exhibits cross-species bioactivity. Both human and rat Recombinant CNTF are synthesized as a 199 amino acid polypeptide (22.7 kDa) lacking a hydrophobic N-terminal signal for secretion.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10072-236
  • 10072-246
  • Protein/peptide name:
  • CNTF
  • CNTF
  • Protein synonyms:
  • HCNTF|CNTF|Ciliary neurotrophic factor
  • CNTF|Ciliary neurotrophic factor
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Rat
  • Source:
  • E. coli
  • E. coli
  • Biological activity:
  • The ED50 was determined by the dose - dependent proliferation of human TF - 1 cells is < 2.0 ng/mL, corresponding to a specific activity of > 5 x 105 units/mg.
  • Biological activity was determined by its ability to stimulate proliferation of human TF - 1 cells using a concentration range of 25.0 - 35.0 ng/mL.
  • Purity:
  • >98%
  • >98%
  • Formulation:
  • Lyophilized
  • Lyophilized

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 0.005 mg
    Log in for availability
    Log in for availability
    Each (5µG)
    Each (5µG) - $510.34
    $510.34
    Each (5µG)
       $510.34
    Minimum order quantity is 1

    Supplier Number: 40-502
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10072-236
    Protein/peptide name
    CNTF
    Protein synonyms
    HCNTF|CNTF|Ciliary neurotrophic factor
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Biological activity
    The ED50 was determined by the dose - dependent proliferation of human TF - 1 cells is < 2.0 ng/mL, corresponding to a specific activity of > 5 x 105 units/mg.
    Purity
    >98%
    Formulation
    Lyophilized
  • 0.005 mg
    Log in for availability
    Log in for availability
    Each (5µG)
    Each (5µG) - $510.34
    $510.34
    Each (5µG)
       $510.34
    Minimum order quantity is 1

    Supplier Number: 40-510
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10072-246
    Protein/peptide name
    CNTF
    Protein synonyms
    CNTF|Ciliary neurotrophic factor
    Protein/peptide type
    Recombinant
    Species
    Rat
    Source
    E. coli
    Biological activity
    Biological activity was determined by its ability to stimulate proliferation of human TF - 1 cells using a concentration range of 25.0 - 35.0 ng/mL.
    Purity
    >98%
    Formulation
    Lyophilized