Order Entry
Canada
ContactUsLinkComponent
Burkholderia pseudomallei Recombinant Areomonas Aminopeptidase (from E. coli)
Burkholderia pseudomallei Recombinant Areomonas Aminopeptidase (from E. coli)
Catalog # 10072-562
Supplier:  Prosci
CAS Number:  
Burkholderia pseudomallei Recombinant Areomonas Aminopeptidase (from E. coli)
Catalog # 10072-562
Supplier:  Prosci
Supplier Number:  40-513
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Burkholderia pseudomallei
  • Size:
    0.1 mg
  • Form:
    Lyophilized
  • Enzyme name:
    Areomonas Aminopeptidase
  • Enzyme activity:
    Sequentially cleaves N-terminal amino acids except E, D, and X-P.
  • Purity:
    >98%
  • Cat. no.:
    10072-562
  • Supplier No.:
    40-513

Specifications

About this item

Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Most proteases act in a specific manner, hydrolyzing bonds at or adjacent to specific residues or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Recombinant Aeromonas Aminopeptidase is a 31.4 kDa protein containing 291 amino acid residues.