Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Kidney Tumor Lysate, ProSci Inc

Tumor, Cryopreserved, 0.1 mg, Human, Kidney
  10062-152
 :   Prosci
 :  1324
Kidney Tumor Lysate, ProSci Inc
  10062-152
 :  Prosci
 :  
Kidney Tumor Lysate, ProSci Inc

Tumor, Cryopreserved, 0.1 mg, Human, Kidney

 :  
 
$736.03
   
Each (100µG)
 
 

Kidney tumor tissue lysate was prepared by homogenization in modified RIPA buffer (150 mM sodium chloride, 50 mM Tris-HCl, pH 7.4, 1 mM ethylenediaminetetraacetic acid, 1 mM phenylmethylsulfonyl fluoride, 1% Triton X-100, 1% sodium deoxycholic acid, 0.1% sodium dodecylsulfate, 5 μg/ml of aprotinin, 5 μg/ml of leupeptin

Tissue and cell debris was removed by centrifugation. Protein concentration was determined with Bio-Rad protein assay. The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 5% β-mercaptoethanol.

 

 

  • Cell Format:
    Cryopreserved
  • Description:
    Tumor
  • Size:
    0.1 mg
  • Source:
    Human
  • Tissue:
    Kidney
  • Cat. No.:
    10062-152
  • Storage conditions:
    Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
  • Concentration:
    2 mg/mL
  • Tested applications:
    WB
  • Storage buffer:
    Kidney Tumor lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.