Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Mouse Adipose Lysate, ProSci

Normal, Cryopreserved, 0.1 mg, Mouse, Adipose
  10062-242
 :   Prosci
 :  1418
Mouse Adipose Lysate, ProSci
  10062-242
 :  Prosci
 :  
Mouse Adipose Lysate, ProSci

Normal, Cryopreserved, 0.1 mg, Mouse, Adipose

 :  
 
$341.68
   
Each (100µG)
 
 

Adipose tissue lysate was prepared by homogenization in modified RIPA buffer (150 mM sodium chloride, 50 mM Tris-HCl, pH 7.4, 1 mM ethylenediaminetetraacetic acid, 1 mM phenylmethylsulfonyl fluoride, 1% Triton X-100, 1% sodium deoxycholic acid, 0.1% sodium dodecylsulfate, 5 μg/ml of aprotinin, 5 μg/ml of leupeptin

Tissue and cell debris was removed by centrifugation. Protein concentration was determined with Bio-Rad protein assay. The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 5% β-mercaptoethanol.

 

 

  • Cell Format:
    Cryopreserved
  • Description:
    Normal
  • Size:
    0.1 mg
  • Source:
    Mouse
  • Tissue:
    Adipose
  • Cat. No.:
    10062-242
  • Storage conditions:
    Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
  • Concentration:
    2 mg/mL
  • Tested applications:
    WB
  • Storage buffer:
    Adipose lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.