Order Entry
Canada
Orders LinkContactUsLinkComponent
Rectum Lysate, ProSci

Rectum Lysate, ProSci

Normal, Cryopreserved, 0.1 mg, Human, Rectum
Catalog # 10062-200
Supplier:   Prosci
Supplier #:  1373
Rectum Lysate, ProSci
Catalog # 10062-200
Supplier:  Prosci
CAS Number:  
Rectum Lysate, ProSci

Normal, Cryopreserved, 0.1 mg, Human, Rectum

CAS Number:  
Price
$332.68
Log in for availability
Each (100µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Rectum tissue lysate was prepared by homogenization in homogenization in lysis buffer (10 mM HEPES pH7.9, 1.5 mM MgCl2, 10 mM KCl, 1 mM ethylenediaminetetraacetic acid, 10% glycerol, 1% NP-40, and a cocktail of protease inhibitors)

Tissue and cell debris was removed by centrifugation. The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 5% β-mercaptoethanol.

Documents

Specifications

Specifications

  • Cell Format:
    Cryopreserved
  • Description:
    Normal
  • Size:
    0.1 mg
  • Source:
    Human
  • Tissue:
    Rectum
  • Cat. No.:
    10062-200
  • Storage conditions:
    Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
  • Concentration:
    2 mg/mL
  • Tested applications:
    WB
  • Storage buffer:
    Rectum lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.