Order Entry
Canada
Orders LinkContactUsLinkComponent
Lymph Node Lysate, ProSci Inc

Lymph Node Lysate, ProSci Inc

Normal, Cryopreserved, 0.1 mg, Human, Lymphnode
Catalog # 10062-192
Supplier:   Prosci
Supplier #:  1369
Lymph Node Lysate, ProSci Inc
Catalog # 10062-192
Supplier:  Prosci
CAS Number:  
Lymph Node Lysate, ProSci Inc

Normal, Cryopreserved, 0.1 mg, Human, Lymphnode

CAS Number:  
Price
332,68 $
Log in for availability
Each (100µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Lymph Node tissue lysate was prepared by homogenization in lysis buffer (10 mM HEPES pH7.9, 1.5 mM MgCl2, 10 mM KCl, 1 mM ethylenediaminetetraacetic acid, 10% glycerol, 1% NP-40, and a cocktail of protease inhibitors)

Tissue and cell debris was removed by centrifugation. The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 5% β-mercaptoethanol.

Documents

Specifications

Specifications

  • Cell Format:
    Cryopreserved
  • Description:
    Normal
  • Size:
    0.1 mg
  • Source:
    Human
  • Tissue:
    Lymphnode
  • Cat. No.:
    10062-192
  • Storage conditions:
    Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
  • Concentration:
    2 mg/mL
  • Tested applications:
    WB
  • Storage buffer:
    Lymph Node lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.