Vikan® Tube Cleaners, 2", Remco Products
Supplier: Remco Products
Certificates
About this item
This tube brush is great for cleaning small pipes and very narrow spaces between machine parts.
- Dishwasher safe
- Autoclavable up to 250 °F
Block material is polypropylene; bristle material is polyester.
The bristles are attached to a helically wound stainless steel shaft which makes it strong and chemically resistant.
Certifications: FDA compliant raw material.
Frequently Bought Together