Rat Recombinant IL2 (from E. coli)
About this item
IL-2 is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions which are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine activated killer cells, monocytes, macrophages and oligodendrocytes. Recombinant murine IL-2 is a 17.2 kDa protein, containing 149 amino acid residues. Recombinant rat IL-2 is a 15.3 kDa protein containing 134 amino acid residues. Recombinant human IL-2 is a 15.5 kDa protein, containing 134 amino acid residues including one intrachain disulfide bond.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
4 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
IL-2 is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions which are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine activated killer cells, monocytes, macrophages and oligodendrocytes. Recombinant murine IL-2 is a 17.2 kDa protein, containing 149 amino acid residues. Recombinant rat IL-2 is a 15.3 kDa protein containing 134 amino acid residues. Recombinant human IL-2 is a 15.5 kDa protein, containing 134 amino acid residues including one intrachain disulfide bond.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10072-422
- 10072-476
- 10072-362
- 10073-086
- Protein/peptide name:
- IL2
- IL2
- IL2
- IL2
- Protein synonyms:
- Interleukin-2|IL-2|Il-2|T-cell growth factor
- Interleukin-2|IL-2|Il-2|T-cell growth factor
- Interleukin-2|TCGF|IL-2|lymphokine|T-cell growth factor
- Interleukin-2|TCGF|IL-2|lymphokine|T-cell growth factor
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Mouse
- Rat
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Biological activity:
- The ED50 was determined by the dose dependent stimulation of murine CTLL - 2 cells is < 0.2 ng/mL, corresponding to a specific activity of > 5 x 106 units/mg.
- The ED50 was determined by the dose - dependent stimulation of the proliferation of murine CTLL - 2 cells is < 0.4 ng/mL, corresponding to a specific activity of > 2.5 x 106 units/mg.
- The ED50 was determined by the dose - dependent stimulation of murine CTLL - 2 cells is ≤ 0.1 ng/mL, corresponding to a specific activity of ≥ 1 x 107 units/mg.
- The ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is = 0.1 ng/mL, corresponding to a specific activity of = 1 x 107 units/mg.
- Purity:
- >98%
- >98%
- >98%
- >98%
- Formulation:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: 40-431Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10072-422Protein/peptide nameIL2Protein synonymsInterleukin-2|IL-2|Il-2|T-cell growth factorProtein/peptide typeRecombinantSpeciesMouseSourceE. coliBiological activityThe ED50 was determined by the dose dependent stimulation of murine CTLL - 2 cells is < 0.2 ng/mL, corresponding to a specific activity of > 5 x 106 units/mg.Purity>98%FormulationLyophilized - Supplier Number: 40-462Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10072-476Protein/peptide nameIL2Protein synonymsInterleukin-2|IL-2|Il-2|T-cell growth factorProtein/peptide typeRecombinantSpeciesRatSourceE. coliBiological activityThe ED50 was determined by the dose - dependent stimulation of the proliferation of murine CTLL - 2 cells is < 0.4 ng/mL, corresponding to a specific activity of > 2.5 x 106 units/mg.Purity>98%FormulationLyophilized - Supplier Number: 40-243Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10072-362Protein/peptide nameIL2Protein synonymsInterleukin-2|TCGF|IL-2|lymphokine|T-cell growth factorProtein/peptide typeRecombinantSpeciesHumanSourceE. coliBiological activityThe ED50 was determined by the dose - dependent stimulation of murine CTLL - 2 cells is ≤ 0.1 ng/mL, corresponding to a specific activity of ≥ 1 x 107 units/mg.Purity>98%FormulationLyophilized - Supplier Number: 40-599Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10073-086Protein/peptide nameIL2Protein synonymsInterleukin-2|TCGF|IL-2|lymphokine|T-cell growth factorProtein/peptide typeRecombinantSpeciesHumanSourceE. coliBiological activityThe ED50 was determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is = 0.1 ng/mL, corresponding to a specific activity of = 1 x 107 units/mg.Purity>98%FormulationLyophilized
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



