Orders LinkContactUsLinkComponent
Plain Microscope Slides, Globe Scientific
Plain Microscope Slides, Globe Scientific
Supplier:   Globe Scientific
Plain Microscope Slides, Globe Scientific
Supplier:  Globe Scientific
Certificates
Plain Microscope Slides, Globe Scientific
Plain Microscope Slides, Globe Scientific
3 Options Available Below
Product Details & Documents
Globe Scientific microscope slides are produced from premium glass sheets and offer exceptional quality while providing considerable savings.
  • Dimensions: 25×75×1 mm
  • Thickness: 1.1 mm
Globe Scientific plain microscope slides are available in three styles. Choose from ground edge, ground edge with safety corners (slightly clipped corners for comfort), or beveled edge with clipped corners. The slides are pre-cleaned and supplied ready for use.
Ordering information: Slides are packaged 72 per box, 20 boxes per case (10 gross).
Product Family Options
Product Information
  • Thickness
    L×W
    Availability
    Price
  • 10118-594
    Plain slides, beveled edges, clipped corners
    Plain slides, beveled edges, clipped corners
    1.1 mm
    75×25 mm (3×1")
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 72
    Pack of 72 - $69.22
    $69.22
    New Customer Promotional Price
    Log in for availability
    Case of 1440
    Case of 1440 - $1,384.47
    $1,384.47
    New Customer Promotional Price
    Pack of 72
       $69.22
    Case of 1440
       $1,384.47
    Supplier Number: 1331
  • 89500-466
    Plain slides, ground edges, safety corners
    Plain slides, ground edges, safety corners
    1.1 mm
    75×25 mm (3×1")
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 72
    Pack of 72 - $62.20
    $62.20
    New Customer Promotional Price
    Log in for availability
    Case of 1440
    Case of 1440 - $1,243.97
    $1,243.97
    New Customer Promotional Price
    Pack of 72
       $62.20
    Case of 1440
       $1,243.97
    Supplier Number: 1321
  • 89500-468
    Plain slides, ground edges, 90° corners
    Plain slides, ground edges, 90° corners
    1.1 mm
    75×25 mm (3×1")
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 72
    Pack of 72 - $63.22
    $63.22
    New Customer Promotional Price
    Log in for availability
    Case of 1440
    Case of 1440 - $1,264.35
    $1,264.35
    New Customer Promotional Price
    Pack of 72
       $63.22
    Case of 1440
       $1,264.35
    Supplier Number: 1301