Orders LinkContactUsLinkComponent
Frosted Microscope Slides, Globe Scientific
Frosted Microscope Slides, Globe Scientific
Supplier:   Globe Scientific
Frosted Microscope Slides, Globe Scientific
Supplier:  Globe Scientific
Certificates
Frosted Microscope Slides, Globe Scientific
Frosted Microscope Slides, Globe Scientific
6 Options Available Below
Product Details & Documents
Globe Scientific microscope slides are produced from premium glass sheets and offer exceptional quality while providing considerable savings.
  • Dimensions: 25×75×1 mm
  • Thickness: 1.1 mm
These frosted microscope slides are produced using a unique chemical etching process. This results in slides with a smooth frosted finish that won't wear out slide marking pens. Choose from single-sided or double-sided frostings, and ground or beveled edges. Ground-edge slides are also available with 90° corners (straight-edge), or safety corners (slightly clipped for comfort). All slides are pre-cleaned and supplied ready for use.
Packaging: Slides are packaged 72 slides per box, 20 boxes per case (10 gross).
Product Family Options
Beveled Edge Microscope Slides
Product Information
  • Thickness
    L×W
    Availability
    Price
  • 10118-596
    Single frosted slides, clipped corners
    Single frosted slides, clipped corners
    1.1 mm
    75×25 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 72
    Pack of 72 - $84.64
    $84.64
    New Customer Promotional Price
    Log in for availability
    Case of 1440
    Case of 1440 - $1,692.78
    $1,692.78
    New Customer Promotional Price
    Pack of 72
       $84.64
    Case of 1440
       $1,692.78
    Supplier Number: 1334
  • 10118-598
    Double frosted slides, clipped corners
    Double frosted slides, clipped corners
    1.1 mm
    75×25 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 72
    Pack of 72 - $88.34
    $88.34
    New Customer Promotional Price
    Log in for availability
    Case of 1440
    Case of 1440 - $1,766.86
    $1,766.86
    New Customer Promotional Price
    Pack of 72
       $88.34
    Case of 1440
       $1,766.86
    Supplier Number: 1338
Ground Edge Microscope Slides
Product Information
  • Thickness
    L×W
    Availability
    Price
  • 89500-470
    Single frosted slides, safety corners
    Single frosted slides, safety corners
    1.1 mm
    75×25 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 72
    Pack of 72 - $74.88
    $74.88
    New Customer Promotional Price
    Log in for availability
    Case of 1440
    Case of 1440 - $1,497.68
    $1,497.68
    New Customer Promotional Price
    Pack of 72
       $74.88
    Case of 1440
       $1,497.68
    Supplier Number: 1324
  • 89500-472
    Single frosted slides, 90° corners
    Single frosted slides, 90° corners
    1.1 mm
    75×25 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 72
    Pack of 72 - $75.55
    $75.55
    New Customer Promotional Price
    Log in for availability
    Case of 1440
    Case of 1440 - $1,510.96
    $1,510.96
    New Customer Promotional Price
    Pack of 72
       $75.55
    Case of 1440
       $1,510.96
    Supplier Number: 1304
  • 89500-474
    Double frosted slides, safety corners
    Double frosted slides, safety corners
    1.1 mm
    75×25 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 72
    Pack of 72 - $57.24
    $57.24
    New Customer Promotional Price
    Log in for availability
    Case of 1440
    Case of 1440 - $1,144.78
    $1,144.78
    New Customer Promotional Price
    Pack of 72
       $57.24
    Case of 1440
       $1,144.78
    Supplier Number: 1328
  • 10118-592
    Double frosted slides, 90° corners
    Double frosted slides, 90° corners
    1.1 mm
    75×25 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 72
    Pack of 72 - $75.33
    $75.33
    New Customer Promotional Price
    Log in for availability
    Case of 1440
    Case of 1440 - $1,506.60
    $1,506.60
    New Customer Promotional Price
    Pack of 72
       $75.33
    Case of 1440
       $1,506.60
    Supplier Number: 1308