Specifications
- Antibody type:Primary
- Antigen name:KIT ligand
- Antigen symbol:KITLG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:0.1 mg
- Form:Lyophilized
- Gene ID:P21583
- Antigen synonyms:KITLG
- Shipping temperature:RT
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hSCF (human SCF).
- Purification:Anti-hSCF specific antibody was purified by affinity chromatography employing immobilized hSCF matrix.
- Cat. no.:10097-580
- Supplier No.:XP-5264
Specifications
About this item
Stem Cell Factor (SCF) is a hematopoietic growth factor that exerts its activity at the early stages of hematopoiesis. SCF stimulates the proliferation of mast cells and is able to augment the proliferation of both myeloid and lymphoid hematopoietic progenitors in bone marrow culture. It also mediates cell to cell adhesion and acts synergistically with other cytokines, probably interleukins. A soluble form is produced by proteolytic processing of isoform 1 in the extracellular domain.
ELISA: To detect hSCF by direct ELISA (using 100 µL/well antibody solution) a concentration of at least 0.5 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant hSCF. Sandwich: To detect hSCF by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5 - 2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Human SCF (XP-5264Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hSCF. Western Blot: To detect hSCF by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hSCF is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
Anti-hSCF specific antibody was purified by affinity chromatography employing immobilized hSCF matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: KITLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human