Anti-EIF4E Rabbit Polyclonal Antibody
Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100, Immunofluorescence: 1:100~1:200
Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: EIF4E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
- Size:0.1 mg
- Western blot:Yes
- Clonality:Polyclonal
- Modification:Ser 209
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
- Antibody type:Primary
- Form:Liquid
- ImmunoFluorescence:Yes
- Storage temperature:Store antibody at –20 °C for up to one year.
- Host:Rabbit
- Concentration:1 mg/mL
- Molecular weight:25 kDa
- Shipping temperature:4 °C
- Antigen name:eukaryotic translation initiation factor 4E
- Cat. No.:10068-496
- Reactivity:Human
- Gene ID:P06730
- Antigen synonyms:EIF4E
- Immunogen:eIF4E (Phospho-Ser209) antibody was raised against a peptide sequence around phosphorylation site of serine 209 (S-G-S (p) -T-T) derived from Human eIF4E.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:EIF4E
- Storage buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



