Specifications
- Antibody type:Primary
- Antigen name:SNVsSgp1 nucleocapsid
- Antigen symbol:SNVsSgp1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Size:0.1 mg
- Form:Liquid
- Gene ID:Q9E1J8
- Antigen synonyms:SNVsSgp1
- Storage buffer:Sin Nombre virus nucleocapsid antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:N/A
- Storage temperature:Sin Nombre Nucleocapsid antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:Sin Nombre virus nucleocapsid antibody was raised against a 19 amino acid peptide near the center of the Sin Nombre virus nucleocapsid. The immunogen is located within amino acids 170 - 220 of Sin Nombre Virus Nucleocapsid.
- Purification:Sin Nombre virus nucleocapsid antibody is affinity chromatography purified via peptide column.
- Cat. no.:10062-066
- Supplier No.:7685
Specifications
About this item
Sin Nombre virus (SNV) is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS). The hantavirus nucleocapsid protein plays several roles in viral replication and assembly, and is the major antigen in humoral responses in humans and mice. Within the bunyviridae family, the nucleocapsid protein also functions as an RNA chaperone facilitating the formation of the stable genomic RNA “panhandle” and is thought to aid in the replication of bunyavirus RNA.
Sin Nombre virus nucleocapsid antibody can detect 10ng Sin Nombre virus nucleocapsid peptide in ELISA at 1 ?g/ml.
Sin Nombre virus nucleocapsid antibody is affinity chromatography purified via peptide column.
Sin Nombre virus nucleocapsid antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SNVsSgp1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus