Anti-APPL1 Goat Polyclonal Antibody
10112-430
: Prosci
- Antibody type:Primary
- Antigen name:adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1
- Antigen symbol:APPL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Rat
- Western blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:Q9UKG1
- Antigen synonyms:APPL1
- Storage buffer:APPL1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:APPL1 antibody was raised against a 14 amino acid synthetic peptide near the internal region of APPL1.
- Purification:APPL1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10112-430
- Supplier No.:45-271
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 80 kDa band observed in rat Aortic Smooth muscle Cells (lane 1), intensity increased after infection with human aPPL1 adenovirus for 48h (lane 2). Calculated MW of 79.7 kDa according to NP_036228.1. The blot was co-labeled with Tubulin-specific antibodies as a loading control. Recommended concentration: 0.3-1 ?g/mL.
APPL1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: APPL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: