To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 20
- Antigen symbol:CCL20
- Clonality:Monoclonal
- Clone:G3_4G10.3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Size:0.5 mg
- Neutralisation:Yes
- Form:Lyophilized
- Gene ID:P78556
- Antigen synonyms:CCL20
- Shipping temperature:RT
- Immunogen:MIP-3 alpha antibody is produced in mice using highly pure (>98%) recombinant human MIP- 3alpha as the immunizing antigen.
- Purification:This IgG1K antibody was purified from cell culture by Protein A affinity chromatography.
- Cat. no.:10097-928
- Supplier No.:38-236
Macrophage Inflammatory Protein 3 alpha is a chemotactic factor that attracts lymphocytes and, slightly, neutrophils, but not monocytes. It inhibits proliferation of myeloid progenitors in colony formation assays. It binds to CCR6.
Was product number XP-5713-M
Neutralization: To yield one-half maximal inhibition [ND50] of the biological activity of mIL-10 (0.5 ng/mL), a concentration of 3.0-5.0 µg/mL of this antibody is required. ELISA: In a sandwich ELISA (assuming 100 µL/well), a concentration of 4.0-8.0 µg/mL of this antibody will detect at least 1600 pg/mL of recombinant murine IL-10 when used with our biotinylated antigen affinity purified anti-murine IL-10 (XP-5162Bt) as the detection antibody at a concentration of approximately 0.5-1.0 µg/mL. Western blot: To detect mIL-10 by Western Blot analysis this antibody can be used at a concentration of 1.0-2.0 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant mIL-10 is 2.0-4.0 ng/lane, under reducing or non-reducing conditions.
This IgG1K antibody was purified from cell culture by Protein A affinity chromatography.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type: Primary
Antigen: CCL20
Clonality: Monoclonal
Clone: G3_4G10.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human