Order Entry
Canada
ContactUsLinkComponent
 
Anti-TIMP1 Rabbit Polyclonal Antibody (Biotin)
  10097-792
 :  Prosci
undefined
Anti-TIMP1 Rabbit Polyclonal Antibody (Biotin)
  10097-792
 :  Prosci
 :  38-216

 

  • Antibody type:
    Primary
  • Antigen name:
    TIMP metallopeptidase inhibitor 1
  • Antigen symbol:
    TIMP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    0.05 mg
  • Form:
    Lyophilized
  • Gene ID:
    P01033
  • Antigen synonyms:
    TIMP1
  • Shipping temperature:
    RT
  • Immunogen:
    Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hTIMP-1 (human Tissue Inhibitor of Metalloproteinases-1). Human TIMP-1 specific antibody was purified by affinity chromatography and then biotinylated.
  • Cat. no.:
    10097-792
  • Supplier No.:
    38-216

 

 

Tissue inhibitor of metalloproteinases 1 (TIMP1) and Tissue inhibitor of metalloproteinases 2 (TIMP 2) have similar properties, specifically in inhibiting enzymes of the matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP1 forms a complex of 1:1 stoichiometry with activated interstitial collagenases, activated stromelysin, active form of 72kDa Type IV collagenase (also known as MMP2 or gelatinase A), and latent and active forms of 92kDa Type IV collagenase (also known as MMP0 or gelatinase B). TIMPs inhibit the proteolytic invasiveness of tumour cells and normal placental trophoblast cells. TIMP1 is produced in low (pg/mL) levels by most cell types. Treatment of cells with the phorbol ester TPA stimulates production of TIMP1 in some cell types, but the low protein levels produced often require concentration of cell culture media to visualize the bands by Western Blotting.

ELISA:
Direct:
To detect hTIMP-1 by direct ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hTIMP-1.

Sandwich
To detect hTIMP-1 by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our polyclonal Anti-Human TIMP-1 as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hTIMP-1.

Western Blot:
To detect hTIMP-1 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hTIMP-1 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: TIMP1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human