Anti-EIF4E2 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
EIF4E2 belongs to the eukaryotic initiation factor 4E family. It recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.
EIF4E2 antibody can be used for detection of EIF4E2 by ELISA at 1:62500. EIF4E2 antibody can be used for detection of EIF4E2 by western blot at 2.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: EIF4E2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
10108-066.pdf
Datasheet
Specifications
- Size:100 ul
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Antibody is purified by protein A chromatography method.
- Antibody type:Primary
- Form:Liquid
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store EIF4E2 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Host:Rabbit
- Molecular weight:27 kDa
- Shipping temperature:4 °C
- Antigen name:eukaryotic translation initiation factor 4E family member 2
- Cat. No.:10108-066
- Reactivity:Human
- Gene ID:O60573
- Antigen synonyms:EIF4E2
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human EIF4E2.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:EIF4E2
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



