Specifications
- Antibody type:Primary
- Antigen name:methylthioribose-1-phosphate isomerase homolog (S. cerevisiae)
- Antigen symbol:Mri1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q9CQT1
- Antigen synonyms:Mri1
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular weight:41 kDa
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store 2410018C20RIK antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human 2410018C20RIK.
- Purification:Antibody is purified by protein A chromatography method.
- Cat. no.:10106-624
- Supplier No.:28-658
Specifications
About this item
This protein was disclosed by the RIKEN Mouse Gene Encyclopaedia Project, a systematic approach to determining the full coding potential of the mouse genome, involves collection and sequencing of full length complementary DNAs and physical mapping of the corresponding genes to the mouse genome.
2410018C20RIK antibody can be used for detection of 2410018C20RIK by ELISA at 1:1562500. 2410018C20RIK antibody can be used for detection of 2410018C20RIK by western blot at 2.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Mri1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse