Orders LinkContactUsLinkComponent
CryoClear™ Cryogenic Vials, External Threads, Globe Scientific
CryoClear™ Cryogenic Vials, External Threads, Globe Scientific
CryoClear™ Cryogenic Vials, External Threads, Globe Scientific
Supplier:  Globe Scientific
Certificates
CryoClear™ Cryogenic Vials, External Threads, Globe Scientific
CryoClear™ Cryogenic Vials, External Threads, Globe Scientific
6 Options Available Below
Product Details & Documents
Innovation to protect precious samples.
  • Non-pyrogenic, non-cytotoxic, non-mutagenic, heavy metals free, BSE/TSE free
  • RNase, DNase, ATP, and Human DNA free
  • Tamper evident packaging
  • Autoclavable
The best and most leak-resistant cryogenic vials available for the long-term storage of your precious samples at temperatures as low as –196 °C.
The vials feature an innovative screw cap that is co-molded with a layer of thermoplastic elastomer (TPE). This special process provides the leak-proof benefits of an O-ring, while eliminating the risk of contamination and sample loss associated with the use of separate O-rings.
Each vial is printed with a unique barcode that may be used for automated data collection, accurate sample inventory or to conceal the sample's identity.
Packaging: Packaged in tamper-evident bags of 50.
Caution: Not for use in the liquid phase of liquid nitrogen.
Product Family Options
Product Information
  • Capacity
    Bottom style
    Cap included
    Sterility
    Availability
    Price
  • 1.00 ml
    Conical bottom, self-standing
    Yes
    Sterile
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $504.83
    $504.83
    New Customer Promotional Price
    Case of 500
       $504.83
    Supplier Number: 3010
    Specifications:
    More
    Specifications
    Cat. No.
    10118-708
  • 2.00 ml
    Round bottom
    Yes
    Sterile
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $408.21
    $408.21
    New Customer Promotional Price
    Case of 500
       $408.21
    Supplier Number: 3011
    Specifications:
    More
    Specifications
    Cat. No.
    10118-710
  • 2.00 ml
    Round bottom, self-standing
    Yes
    Sterile
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $504.83
    $504.83
    New Customer Promotional Price
    Case of 500
       $504.83
    Supplier Number: 3012
    Specifications:
    More
    Specifications
    Cat. No.
    10118-712
  • 3.00 ml
    Round bottom
    Yes
    Sterile
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $623.59
    $623.59
    New Customer Promotional Price
    Case of 500
       $623.59
    Supplier Number: 3013
    Specifications:
    More
    Specifications
    Cat. No.
    10118-714
  • 4.00 ml
    Round bottom, self-standing
    Yes
    Sterile
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $631.18
    $631.18
    New Customer Promotional Price
    Case of 500
       $631.18
    Supplier Number: 3014
    Specifications:
    More
    Specifications
    Cat. No.
    10118-716
  • 5.00 ml
    Round bottom, self-standing
    Yes
    Sterile
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $681.34
    $681.34
    New Customer Promotional Price
    Case of 500
       $681.34
    Supplier Number: 3015
    Specifications:
    More
    Specifications
    Cat. No.
    10118-718