Dura-Clamp, Dynalon
Catalog #: 62849-112
Supplier: Dynalon
Specifications
- Cat. no.:62849-112
- L×W×H:1 x 1/3 x 2/3"
- Supplier No.:670715
Specifications
About this item
Polypropylene Dura-Clamp tubing flow valve is easily operated with one hand and will not rust or corrode
Accepts tubing O.D. up to 1/4".
Customers Who Bought This Also Bought