ContactUsLinkComponent
Gastight® CTC PAL® S-Line Syringes for HPLC Autosamplers, Hamilton
Gastight® CTC PAL® S-Line Syringes for HPLC Autosamplers, Hamilton
Supplier:  Hamilton
Certificates
undefined
Gastight® CTC PAL® S-Line Syringes for HPLC Autosamplers, Hamilton
Supplier:  Hamilton
Specifications
  • Catalog No:
  • 89498-976
  • 89498-978
  • 89498-980
  • 89498-982
  • 89498-972
  • 89498-974
  • Type:
  • Gastight
  • Gastight
  • Gastight
  • Gastight
  • Gastight
  • Gastight
  • Series:
  • 1700
  • 1700
  • 1700
  • 1700
  • 1700
  • 1700
  • Autoclavable:
  • No
  • No
  • No
  • No
  • No
  • No
  • Ext.Ø:
  • 0.711 mm (Needle)
  • 0.711 mm (Needle)
  • 0.711 mm (Needle)
  • 0.711 mm (Needle)
  • 0.711 mm (Needle)
  • 0.711 mm (Needle)
  • Needle length:
  • 51 mm
  • 51 mm
  • 51 mm
  • 51 mm
  • 51 mm
  • 51 mm
  • Point style:
  • 3
  • 3
  • 3
  • 3
  • 3
  • 3
  • Termination:
  • Cemented Needle (N)
  • Cemented Needle (N)
  • Cemented Needle (N)
  • Cemented Needle (N)
  • Cemented Needle (N)
  • Cemented Needle (N)