ContactUsLinkComponent
Beakers, Square Ratio, Polypropylene, Dynalon
Beakers, Square Ratio, Polypropylene, Dynalon
Catalog # 30620-146
Supplier:  Dynalon
Supplier #: 522085-0100
undefined
Beakers, Square Ratio, Polypropylene, Dynalon
Catalog # 30620-146
Supplier:  Dynalon
Supplier Number:  522085-2000
Supplier #: 522085-0100
Log in for availability
Select an Option
60 mm (Top)
Select an Option
10 ml
Select an Option
100 ml (3.4 oz.)
Select an Option
30620-146
Select an Option
0.03937 lbs.
Select an Option
522085-0100
About this item
Square ratio beakers are a safe, convenient alternative to griffin style beakers
These beakers feature a vertical lip around the top of the beaker which serves two purposes: first the lip contains many spills that may occur with the rounded design of a griffin style beaker. Second, this vertical lip allows the beakers to be stacked together in sets when not in use. Maximum use temperature is 121°C.
Frequently Bought Together