Order Entry
Canada
ContactUsLinkComponent
Stir Bar Retriever, Polypropylene, Dynalon
Stir Bar Retriever, Polypropylene, Dynalon
Catalog # 30620-472
Supplier:  Dynalon
undefined
Stir Bar Retriever, Polypropylene, Dynalon
Catalog # 30620-472
Supplier:  Dynalon
Supplier Number:  303325

Specifications

  • Ø:
    10 mm (3/8")
  • Description:
    Weight: 0.05375 lbs.
  • Length:
    305 mm (12")
  • Cat. no.:
    30620-472
  • Supplier No.:
    303325

Specifications

About this item

The polypropylene stir bar retriever can be used with all stir bars

Polypropylene is chemically resistant and may be used up to 135 °C.

Delivery: Includes hanging ring for storage convenience.
Packaging: Weight: 0.05375 lbs.