Stir Bar, Extra Large, PTFE, Dynalon
Supplier: Dynalon
Jump to:
About this item
Extra large polygonal stir bar for use in very large containers
Powerful permanent Alnico V magnetic core. PTFE has excellent chemical resistance and can be used up to 260°C. Always store in pairs to maintain magnetic strength.
3 Options Available Below
Product Details & Documents
Extra large polygonal stir bar for use in very large containers
Powerful permanent Alnico V magnetic core. PTFE has excellent chemical resistance and can be used up to 260°C. Always store in pairs to maintain magnetic strength.
Product Family Options
Product Information
- ShapeLengthØCoatingWeightAvailabilityPrice
- Supplier Number: 304445-0001
- Supplier Number: 304445-0002
- Supplier Number: 304445-0003