ContactUsLinkComponent
Nalgene® Biotainer Replacement Closures, Thermo Scientific
Nalgene® Biotainer Replacement Closures, Thermo Scientific
  89429-586
Nalgene® Biotainer Replacement Closures, Thermo Scientific
  89429-586
 :  362515-0480
  • Cap Size:
    48 mm
  • Description:
    Biotainer Replacement Closures
  • Cat. no.:
    89429-586
  • Supplier No.:
    362515-0480
Polypropylene Biotainer Replacement Closures are designed to protect the contents within InVitro™ Biotainer™ bottles and carboys
Closures feature silicone liners and are compatable with 1L, 2L, 5L, 10L, and 20L polycarbonate Biotainers. Closure size is 48mm. Sterile.