Order Entry
ContactUsLinkComponent
Anti-IgG Rabbit Polyclonal Antibody (Texas Red®)
Anti-IgG Rabbit Polyclonal Antibody (Texas Red®)
Catalog #: RL706-409-002
Anti-IgG Rabbit Polyclonal Antibody (Texas Red®)
Catalog #: RL706-409-002
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Guinea pig
  • Antigen symbol:
    IgG
  • Conjugation:
    Texas Red®
  • ImmunoFluorescence:
    Yes
  • Size:
    500 µL
  • Epitope:
    Heavy and Light chain
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Antigen synonyms:
    F(ab')2 Anti-Guinea Pig IgG (H&L)
  • Antibody fragment:
    F(ab')2
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Guinea Pig IgG whole molecule
  • Tested applications:
    Fluoresence ELISA, Fluoresence Western Blot
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. no.:
    RL706-409-002
  • Supplier No.:
    706-409-002

Specifications

About this item

Recommended Dilutions:
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Guinea Pig

Customers Who Bought This Also Bought