Order Entry
ContactUsLinkComponent
Anti-Biotin Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anti-Biotin Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog #: RL600-405-098
Anti-Biotin Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog #: RL600-405-098
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Biotin
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Antigen symbol:
    Biotin
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 0.1% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
  • Storage temperature:
    +4 °C
  • Concentration:
    0.5 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Wet Ice
  • Immunogen:
    Biotin conjugated to Keyhole Limpet Hemocyanin (KLH)
  • Tested applications:
    Dot Blot
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. no.:
    RL600-405-098
  • Supplier No.:
    600-405-098

Specifications

About this item

Suitable for immunoblotting (western or dot blot), ELISA and immunochemistry as well as other phosphatase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Customers Who Bought This Also Bought