To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:Actin Beta
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Hamster,Pig,Monkey,Sheep,Goat,Human,Bovine,Guinea pig,Rat,Mouse
- Antigen symbol:ACTB
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen synonyms:PS1TP5-binding protein 1,cytoplasmic 1,PS1TP5BP1,BRWS1,beta cytoskeletal actin,actin
- Amino acid number:359 to 368
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Beta Actin Antibody Peroxidase Conjugated antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 359-368 of Human beta Actin.
- Purification:affinity-purified
- Cat. no.:RL600-403-886
- Supplier No.:600-403-886
Specifications
About this item
Beta Actin Antibody Peroxidase Conjugated antibody has been tested for use in ELISA, immunochemistry and western blot.
Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Type: Control
Antigen: ACTB
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human, Monkey, Sheep, Bovine, Goat, Guinea Pig, Pig, Hamster