Order Entry
ContactUsLinkComponent
Anti-Biotin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-Biotin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog #: RL600-402-098
FITC ANTI-BIOTIN 100UG
Anti-Biotin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog #: RL600-402-098
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Biotin
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Antigen symbol:
    Biotin
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
  • Storage temperature:
    –20 °C
  • Concentration:
    1.0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ambient
  • Immunogen:
    Biotin conjugated to Keyhole Limpet Hemocyanin (KLH)
  • Tested applications:
    Fluoresence ELISA, Fluoresence Western Blot
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. no.:
    RL600-402-098
  • Supplier No.:
    600-402-098

Specifications

About this item

Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: Immunofluorescence: 1:500-1:2500
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Customers Who Bought This Also Bought