To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 4
- Clonality:Polyclonal
- Gene ID:396668
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pig
- Antigen symbol:CCL4
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen synonyms:MIP1b,SCY4A,MIP-1BETA
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:This Protein-A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with recombinant Protein raised in yeast, corresponding to the 69 amino acids of the mature swine CCL4/MIP1beta Protein.
- Purification:Protein A purified
- Cat. no.:RL214-401-B85
- Supplier No.:214-401-B85
Specifications
About this item
This Protein-A purified antibody is suitable for ELISA and Western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 7.8 kDa in size corresponding to swine CCL4 Protein by western blotting.
Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:500;
Type: Primary
Antigen: CCL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig