To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Growth Differentiation Factor 15
- Clonality:Polyclonal
- Gene ID:23886
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:GDF15
- Conjugation:Biotin
- ELISA:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen synonyms:MIC-1,GDF-15,NAG-1,PTGFB,PDF,PLAB,MIC1
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:This Protein-A purified antibody was prepared by repeated immunizations with an MBP-tagged recombinant Protein produced in E.coli corresponding to full-length mouse NAG-1 Protein. Cross reactivity to MBP was removed via cross-adsorption chromatography.
- Purification:purified by immunoaffinity chromatography
- Cat. no.:RL210-406-B68
- Supplier No.:210-406-B68
Specifications
About this item
This Protein-A purified, MBP-cross-adsorbed antibody is suitable for ELISA and western blotting of mouse and human NAG-1 Protein. For detection of NAG-1 in mouse serum, a sandwich ELISA is suggested using this antibody in combination with anti-NAG-1/GDF1.
Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: Gdf15
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse