To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 20
- Clonality:Polyclonal
- Gene ID:20297
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Antigen symbol:CCL20
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen synonyms:MIP3A,exodus-1,Scya20,CKb4,MIP-3[a],ST38,MIP-3A,LARC
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:MIP-3 alpha Antibody Peroxidase Conjugated was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant mouse MIP-3a Protein.
- Cat. no.:RL210-403-B51
- Supplier No.:210-403-B51
Specifications
About this item
This purified antibody has been tested for use in ELISA and western blotting. By western blot a band approximately 10.7 kDa in size corresponding to mouse MIP-3a Protein is expected in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:2500
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!
Type: Primary
Antigen: CCL20
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Mouse