To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 32
- Clonality:Polyclonal
- Gene ID:9235
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:IL32
- Conjugation:Biotin
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen synonyms:TAIFd,IL-32,IL-32alpha,Interleukin-32,TAIFa,TAIFc,IL-32beta,IL-32gamma,NK4,TAIF,Natural killer cells protein 4,TAIFb,IL-32delta
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant human IL-32A Protein.
- Purification:purified
- Cat. no.:RL209-406-B49
- Supplier No.:209-406-B49
Specifications
About this item
This purified antibody has been tested for use in ELISA and western blotting. By western blot a band approximately 15 kDa in size corresponding to native human IL-32a Protein is expected in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: IL32
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human