Order Entry
ContactUsLinkComponent
Anti-IL6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IL6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog #: RL209-403-310
HRP ANTI-HUMAN IL-6 100UG
Anti-IL6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog #: RL209-403-310
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 6
  • Clonality:
    Polyclonal
  • Gene ID:
    3569
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    IL6
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Antigen synonyms:
    HSF,
    HGF,
    IFNB2,
    interferon beta 2,
    BSF2,
    IL-6
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
  • Storage temperature:
    –20 °C
  • Concentration:
    1.1 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ambient
  • Immunogen:
    This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant human IL-6 produced in E.coli.
  • Purification:
    purified
  • Cat. no.:
    RL209-403-310
  • Supplier No.:
    209-403-310

Specifications

About this item

This purified antibody has been tested for use in ELISA and western blotting. Reactivity is also expected in neutralizations, radioimmunoassay and immunochemistry. The endotoxin content is estimated to be <10 pg/µl by the LAL method.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:2500
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought