To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 17 A
- Clonality:Polyclonal
- Gene ID:3605
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:IL17A
- Conjugation:Unconjugated
- ELISA:Yes
- Size:25 µL
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:IL-17,IL17,CTLA8,IL-17A,CTLA-8,Cytotoxic T-lymphocyte-associated serine esterase 8
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.1% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:IL-17A Antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant human IL17-A Protein.
- Purification:affinity-purified
- Cat. no.:RL209-401-B32S
- Supplier No.:209-401-B32S
Specifications
About this item
IL-17-A antibody has been tested for use in ELISA and western blotting. By western blot a band approximately ~17.5 kDa in size corresponding to human IL-17-A Protein is expected in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:1:000 - 1:5000; Western Blot: 1:500 - 1:2000
Type: Primary
Antigen: IL17A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human