To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Colony Stimulating Factor 2
- Clonality:Polyclonal
- Gene ID:1437
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:CSF2
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen synonyms:GMCSF,TUBB2,Csfgm,Gm-CSf,TUBB,MGI-IGM,CDCBM5
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2,
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant human GM-CSF Protein.
- Purification:purified
- Cat. no.:RL209-401-314
- Supplier No.:209-401-314
Specifications
About this item
This purified antibody has been tested for use in neutralizations, ELISA and western blotting. Reactivity is also expected in radioimmunoassay.
Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: CSF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: