About this item
This Protein-A purified antibody is suitable for ELISA and Western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 9.3 kDa in size corresponding to bovine CXCL10 Protein by western blotting.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1 µg/ml;
Type: Primary
Antigen: CXCL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- RL201-401-B84S
- RL201-401-B84
- Antigen name:
- Chemokine (C-X-C motif) Ligand 10
- Chemokine (C-X-C motif) Ligand 10
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Sheep, Goat, Bovine
- Sheep, Goat, Bovine
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 615107
- 615107
- Isotype:
- IgG
- IgG