Order Entry
ContactUsLinkComponent
Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 649) [clone: 18F3.H11]
Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 649) [clone: 18F3.H11]
Catalog #: RL200-343-269
DYLIGHT649 ANTI-AKT PT308
Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 649) [clone: 18F3.H11]
Catalog #: RL200-343-269
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AKT Serine/threonine Kinase 1
  • Clonality:
    Monoclonal
  • Clone:
    18F3.H11
  • Gene ID:
    207
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Antigen symbol:
    AKT1
  • Conjugation:
    DyLight® 649
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Antigen synonyms:
    CWS6,
    RAC,
    RAC-ALPHA,
    PRKBA,
    PKB,
    AKT,
    PKB-ALPHA
  • Modification:
    phospho-Thr308
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
  • Storage temperature:
    –20 °C
  • Concentration:
    1.0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ambient
  • Immunogen:
    This monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding T308 of human AKT1 Protein.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    Protein A purified
  • Cat. no.:
    RL200-343-269
  • Supplier No.:
    200-343-269

Specifications

About this item

This monoclonal antibody is suitable for immunofluorescence microscopy, FLISA and western blotting. Expect a band approximately 56 kDa in size corresponding to phosphorylated AKT Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:4000-1:20000; FLISA: >1:20000; Western Blot: 1:10000 - 1:25000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 18F3.H11
Conjugation: DyLight® 649
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Monkey

Customers Who Bought This Also Bought