To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Transient Receptor Potential Cation Channel Subfamily M member 7
- Clonality:Monoclonal
- Clone:S74-25
- Gene ID:58800
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TRPM7
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:TRPPLIK,CHAK1,LTrpC-7,2310022G15Rik,Ltrpc7,Ltpr7,CHAK,4833414K03Rik,5033407O22Rik
- Storage buffer:0.09% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:TRPM7 Antibody was produced in mice by repeated immunizations raised against a synthetic peptide corresponding to the c-terminus region of mouse TrpM7.
- Purification:Protein G purified
- Cat. no.:RL200-301-G57
- Supplier No.:200-301-G57
Specifications
About this item
Anti-TRPM7 Antibody is suitable for use in WB, IP, and Immunofluorescence microscopy. Antibody is provided in PBS pH 7.4. Expect a band approximately ~220kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions:
Type: Primary
Antigen: TRPM7
Clonality: Monoclonal
Clone: S74-25
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Mouse