To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Transient Receptor Potential Cation Channel Subfamily C member 5
- Clonality:Monoclonal
- Clone:S67-15
- Gene ID:7224
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TRPC5
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:Htrp-5,Htrp5,Short transient receptor potential channel 5,TRP5,TRPC5
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.250% (v/v) Glycerol
- Storage temperature:–20 °C
- Concentration:1mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:TrpC5 Antibody was produced in mice by repeated immunizations raised against a synthetic peptide corresponding to a near c-terminus region of rat TrpC5 (also known as short transient receptor potential channel 5, and Htrp5).
- Purification:Protein G purified
- Cat. no.:RL200-301-G55
- Supplier No.:200-301-G55
Specifications
About this item
Anti-TrpC5 Antibody is suitable for use in WB, Immunochemistryand IP. Expect a band approximately ~110kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions:
Type: Primary
Antigen: TRPC5
Clonality: Monoclonal
Clone: S67-15
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat