To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Potassium Channel Subfamily T Member 1
- Clonality:Monoclonal
- Clone:S3-26
- Gene ID:60444
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Antigen symbol:KCNT1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:near the C-terminus
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:Slack,rSlo2,KIAA1422
- Storage buffer:50% (v/v) Glycerol
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:SLACK potassium channel Antibody was produced in mice by repeated immunizations raised against a fusion Protein near the c-terminus region of rat Slo2.2 (Slack).
- Purification:Protein G purified
- Cat. no.:RL200-301-G46
- Supplier No.:200-301-G46
Specifications
About this item
Anti-SLACK Antibody is suitable for use in WB and IHC. Expect a band approximately ~140kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user. Antibody is provided in PBS pH 7.4.
Recommended Dilutions: Immunofluorescence: 1.0-10 µg/ml; Western Blot: 1 µg/ml; Immunochemistry: 0.1-1.0 µg/ml
Type: Primary
Antigen: KCNT1
Clonality: Monoclonal
Clone: S3-26
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat