To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:SH3 And Multiple Ankyrin Repeat Domains 2
- Clonality:Monoclonal
- Clone:S23b-6
- Gene ID:171093
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SHANK2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:Center
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:AUTS17,CORTBP1,ProSAP1,SHANK,CTTNBP1,SPANK-3
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.250% (v/v) Glycerol
- Storage temperature:–20 °C
- Concentration:1mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:SHANK2 Antibody was produced in mice by repeated immunizations raised against a fusion Protein corresponding to an internal region of (SH3/PDZ domains) of rat Shank2 (SH3 and multiple ankyrin repeat domains Protein 2.
- Purification:Protein G purified
- Cat. no.:RL200-301-G44
- Supplier No.:200-301-G44
Specifications
About this item
Anti-SHANK2 Antibody is suitable for use in WB, Immunochemistryand IP. Expect a band approximately ~160kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions:
Type: Primary
Antigen: SHANK2
Clonality: Monoclonal
Clone: S23b-6
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human, Rat