To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Potassium voltage-gated channel subfamily B member 1
- Clonality:Monoclonal
- Clone:S39-25
- Gene ID:25736
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Antigen symbol:KCNB1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:Center
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:DRK1,h-DRK1,EIEE26,KV2.1
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.250% (v/v) Glycerol
- Storage temperature:–20 °C
- Concentration:1mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:Kv2.1 Extracellular Antibody was produced in mice by repeated immunizations raised against a synthetic peptide corresponding to an internal region (extracellular S1-S2 loop) of rat Kv2.1.
- Purification:Protein G purified
- Cat. no.:RL200-301-F99
- Supplier No.:200-301-F99
Specifications
About this item
Anti-Kv2.1 Extracellular Antibody is suitable for use in WB and Immunofluorescence microscopy. Expect a band approximately ~105-125kDa on specific lysates (varies with cell background due to phosphorylation).
Recommended Dilutions: Immunofluorescence: 1.0-10µg/ml; Western Blot: 1µg/ml; Immunochemistry: 0.1-1.0µg/ml
Type: Primary
Antigen: KCNB1
Clonality: Monoclonal
Clone: S39-25
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat