About this item
This Protein A purified antibody is suitable for ELISA and western blotting of human NAG-1 Protein. For detection of NAG-1 in human serum, a sandwich ELISA is suggested using this antibody in combination with anti-NAG-1/GDF15 C-terminal specific antibodies.
Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:1000;
Type: Primary
Antigen: NAG-1
Clonality: Monoclonal
Clone: 23G10.F8
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- RL200-301-B10S
- RL200-301-B10
- Antigen name:
- Growth Differentiation Factor 15
- Growth Differentiation Factor 15
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 23G10.F8
- 23G10.F8
- Host:
- Mouse
- Mouse
- Gene ID:
- 9518
- 9518
- Isotype:
- IgG1 kappa
- IgG1 kappa