To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:AKT Serine/threonine Kinase 1
- Clonality:Monoclonal
- Clone:14E5.16C8.25F6
- Gene ID:207
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chimpanzee,Human,Rat,Mouse
- Antigen symbol:AKT1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:25 µL
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:CWS6,RAC,RAC-ALPHA,PRKBA,PKB,AKT,PKB-ALPHA
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:Anti-AKT monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues near the C terminal end of of human AKT1 Protein.
- Purification:Protein A purified
- Cat. no.:RL200-301-401S
- Supplier No.:200-301-401S
Specifications
About this item
Anti-AKT Antibody is suitable for ELISA, immunochemistry, immunoprecipitation and western blotting. Expect a band approximately 54 - 56 kDa in size corresponding to AKT Protein by western blotting in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500 - 1:3000; Immunochemistry: 20 µg/ml;
Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 14E5.16C8.25F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chimpanzee