LEGEND MAX™ Mouse IL-34 ELISA Kit with Pre-coated Plates, BioLegend
Catalog # CA439107-BL
Supplier: Biolegend
CAS Number:
Specifications
- Assay Type:Sandwich
- Conjugate ELISA:Avidin-HRP D
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Interleukin 34 (IL-34)
- Description:LEGEND MAX™ Mouse IL-34 ELISA Kit
- Size:1 Kit
- Detection range:31.3 - 2000 pg/mL
- Sensitivity:8.4 ± 1.8 pg/ml
- Cat. no.:CA439107-BL
- Supplier No.:439107
- No. of Tests:1× 96-well strip microplate