Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Protein Linking and Labeling Systems
Cyto-Last™ Buffer, BioLegend
Cyto-Last™ Buffer, BioLegend
Cyto-Last™ Buffer, BioLegend
Catalog # CA422501-BL
Supplier:
Biolegend
CAS Number:
Expand image
Cyto-Last™ Buffer, BioLegend
Catalog # CA422501-BL
Supplier:
Biolegend
Supplier Number:
422501
CAS Number:
Specifications
Description:
Cyto-Last™ Buffer
Size:
100 ml
Flow cytometry:
Yes
Antigen symbol:
CYTO-LAST BUFFER
Cat. no.:
CA422501-BL
Supplier No.:
422501
Specifications
About this item
Product Details
Product Details