About this item
IBI's Midi Fast-Ion Plasmid Kits are endotoxin free and use pre-packed ion-exchange resin columns to purify plasmid or cosmid DNA from bacterial cultures
- Sample Size: 50 ml high-copy plasmid/100 ml low-copy plasmid
- Operation: Gravity Flow Column
- Expectant Yield: 200 µg of plasmid DNA
- Operation Time: 120 minutes or less
The Midi Fast Ion Plasmid Kit uses pre-packed ion-exchange resin columns to purify plasmid or cosmid DNA from bacterial cultures. In this process, the modified alkaline lysis method and RNase treatment are used for creating cleared lysate with minimal genomic DNA and RNA contaminants. Using a gravity-flow procedure, the plasmid DNA in crude lysate has been bound to the column. The contaminants can be washed off with a wash buffer. Finally, the purified DNA is eluted by a high salt buffer and then precipitated with isopropanol for desalting. The entire procedure can be completed in less than two hours. The purified plasmid DNA is suitable for transfection, sequencing reactions, PCR, and in-vitro transcription.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (anion exchange)
- Prep size:Midi
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:Midi fast-ion plasmid sample kit (endotoxin free)
- Yield:Up to 200 µg of plasmid DNA