ContactUsLinkComponent
Corning® PureCoat™ Collagen I, Rectangular Straight Neck Cell Culture Multi-Flask, 3-Layer with Vented Cap, Corning
 :  Corning
undefined
Corning® PureCoat™ Collagen I, Rectangular Straight Neck Cell Culture Multi-Flask, 3-Layer with Vented Cap, Corning
 :  Corning
  • Storage conditions:
    Room temperature for 18 months from date of manufacture
  • Sterility:
    Sterile, Nonpyrogenic
Frequently Bought Together