About this item
Eppendorf® sets the new standard for deep well plates with new RecoverMax well designs.
- Made of virgin polypropylene, each plate type is available as standard and sterile quality
- Ease of stacking, multiple sealing options as well as high chemical and temperature resistance make these plates excellent, multipurpose tools
The well geometry enables high-speed pipetting, excellent mixing and high-speed centrifugation to accelerate your manual as well as your automated work processes. Sophisticated colors and coding make reading, orientation and manual handling easier and faster.
Deep well Plate 96/500, 96/1000 and 384/200 are also available as DNA LoBind and Protein LoBind quality. Reliable centrifugation stability Eppendorf deep well plates 96 and 384 are designed to be centrifugation-stable and durable in a temperature range from 4 to 40 °C. Standard, DNA and Protein LoBind Eppendorf deep well Plate 96 and 384 can be centrifuged up to 6000×g². Sterile Eppendorf plate deep well 96 and 384 are centrifugation stable up to 5000×g².
For sample recovery after processing or storage, centrifuge 5804 R and 5810 R and rotors A-4-81-MTP and A-2-DWP are recommended.
Specifications
- Catalog No:
- CA89208-776
- CA89085-632
- CA89085-620
- CA89085-642
- CA89085-618
- CA89085-640
- CA89085-624
- CA89087-640
- CA89085-646
- CA89085-622
- CA89085-644
- CA89085-628
- CA89085-626
- CA89085-648
- CA89085-630
- CA89087-608
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001